missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Nisin

Product Code. 11495041 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11495041

Brand: MP Biomedicals™ 0215583905

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Nisin content: 2.5% Remainder is NaCl and milk solids (denatured)

  • Molecular Formula: C14H230N42O37S7
  • Molecular Weight: 3354.3

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15° to 30°C
Form Powder
Product Type Antimicrobial
pH 3.31
Quantity 5 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.