missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Nigrosin Stain

Product Code. 11454992 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454992 100 g 100g
11444992 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11454992 Supplier MP Biomedicals™ Supplier No. 0219459380

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as a viable cell staining agent especially in stem cell biology

  • Formula Weight: 737.87
  • CAS # 8005-03-6
  • Synonym: Acid Black 2; C.I. 50420
  • Physical Description: Black crystalline powder
  • Solublility: Soluble in water(1mg/ml-opaque black with purple cast solution)
TRUSTED_SUSTAINABILITY

Specifications

Type Nigrosin
Quantity 100 g
Format Crystal
For Use With (Application) Viable cell staining agent especially in stem cell biology
Storage Requirements 15 to 30°C

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.