missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Min. Ess. Med. Eagle Non-Ess. Amino Acids

Product Code. 11404805 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404805 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11404805 Supplier MP Biomedicals™ Supplier No. 091681049

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For use in cell culture applications using defined media for the in vitro cultivation of cells

TRUSTED_SUSTAINABILITY

Specifications

Product Type Amino Acid Supplement
Sterility Sterile
Without Additives No Glutamine
Quantity 100 mL
pH 2.8 to 3.8
Content And Storage 4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.