missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ MERmaid™ SPIN Kit
Shop All MP Biomedicals Products
Click to view available options
No. of Reactions:
25 Preps
Unit Size:
25 preps
Description
- The MERmaid SPIN Kit is designed for purifying small MW DNA away from gel material, nucleotides, salts, proteins, or other undesirable components of a mixture.
- Single-stranded or double-stranded DNA oligomers or PCR products of 10-200 nucleotides in length are readily purified from agarose and polyacrylamide gels or from solutions.
- Purified DNA can be used in enzymatic reactions, conventional or automated sequencing, cloning and microarray analysis.
Specifications
Specifications
| Format | Kit |
| Sample Type | Enzymatic Reactions, Solutions, Agarose Gels, Polyacrylamide Gels |
| For Use With (Application) | Rapid purification of single or double-stranded DNA from gel material, nucleotides, salts, proteins or other undesirable components of mixture |
| Includes | Glassfog™ SPIN Bind solution, MERmaid™ SPIN high salt wash, Ethanol Wash concentrate, Elution solution, 50X Electrophoresis Buffer, BioGel, MERmaid™ SPIN filters, Elution catch tubes, label block |
| No. of Reactions | 25 Preps |
| Sample Volume | Up to ∼300 mg Gel Slice or up to ∼300 μL DNA Solution |
| Sample Source | Gels and Solutions |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction