missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ M9 Minimal Medium Salts (Maniatis Formulation), Powdered
Sfoglia Tutto MP Biomedicals Prodotti
Click to view available options
Quantity:
227 g
1000 g
Dimensione della confezione:
1kg
227g
Descrizione
- A commonly used base for preparing a defined synthetic medium for the growth of most E. coli host cells
- Powder is reconstituted in sterile purified water, (or water with melted purified agar added)
- Additional nutrients, such as glucose or other carbon source, casamino acids, vitamins, or antibiotics are added when the solution cools
- Add additional nutrients before plates are poured or media is inoculated with bacteria
Specifica
Specifica
| Certifications/Compliance | Molecular Biology Certified |
| Concentration | 56.4 g/L of a 5X Solution |
| Content And Storage | 15° to 30°C |
| Description | A 56.4 g sample of M9 Minimal Salts was dissolved in an aqueous solution to 1 liter, the pH adjusted to 7.4 (if needed), and the solution autoclaved to make a sterile 5X concentrate. |
| Form | Powder |
| Media Type | M9 Minimal Medium |
| Product Type | Microbiology Media |
| Sterility | Sterile |
| Formulation | 33.9 g Na2HPO4; 15 g KH2PO4; 2.5 g NaCl; 5 g NH4Cl |
| For Use With (Application) | Preparing a Defined Synthetic Medium for The Growth of E. Coli Host |
| Vedi altri risultati |
Sicurezza e movimentazione
Identificativo prodotto
- M9 Minimal Medium Salts
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501-Dispose of contents/container to in accordance with all local, regional,national and international regulations.
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto