missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Lysing Matrix H

Product Code. 15444683 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 x 2 mL Tubes
50 x 2 mL Tubes
500 x 2 mL Tubes
Unit Size:
Pack of 100
Pack of 50
Pack of 500
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15444683 500 x 2 mL Tubes Pack of 500
11422430 50 x 2 mL Tubes Pack of 50
11432430 100 x 2 mL Tubes Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15444683 Supplier MP Biomedicals™ Supplier No. 116917500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lysing Matrix H used to isolate RNA and Proteins from bacterial aggregates, biofilms, and whole insects.

  • Impact-resistant 2 mL tube contains 2 mm glass beads and 2 mm yellow zirconium oxide beads
  • With yellow caps
  • Specific Gravity: Glass Beads, 2 mm; Yellow Zirconium Oxide Beads, 5.5
  • Applications: Nucleic Acid Isolation and Purification
TRUSTED_SUSTAINABILITY

Specifications

Bead Type Glass, Yellow Zirconium Oxide
Capacity 2 mL
Diameter (English) 0.068 in.
Diameter (Metric) 2 mL
Sample Type Seeds, Animal Tissues, Plants, Soil, Bacteria, Ancient and Dried Samples, Clay, Wood
DNase Dnase-free
For Use With (Application) Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis
Matrix Lysing matrix H
Quantity 500 x 2 mL Tubes
RNase RNase-free

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.