missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Impact-resistant 2 mL tube contains 2 mm glass beads and 2 mm yellow zirconium oxide beads
- With yellow caps
- Specific Gravity: Glass Beads, 2 mm; Yellow Zirconium Oxide Beads, 5.5
- Applications: Nucleic Acid Isolation and Purification
Specifications
Specifications
| Bead Type | Glass, Yellow Zirconium Oxide |
| Capacity | 2 mL |
| Diameter (English) | 0.068 in. |
| Diameter (Metric) | 2 mL |
| Sample Type | Seeds, Animal Tissues, Plants, Soil, Bacteria, Ancient and Dried Samples, Clay, Wood |
| DNase | Dnase-free |
| For Use With (Application) | Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis |
| Matrix | Lysing matrix H |
| Quantity | 100 x 2 mL Tubes |
| RNase | RNase-free |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?