missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Lysing Matrix F, 2 mL Tubes

Product Code. 11482420 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 x 2 mL Tubes
50 x 2 mL Tubes
500 x 2 mL Tubes
Unit Size:
Each
Pack of 100
Pack of 50
This item is not returnable. View return policy

Product Code. 11482420

Brand: MP Biomedicals™ 116915050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lysing Matrix F, 2 mL Tubes are used to isolate RNA and protein from samples. Lysing Matrix F is useful in applications such as isolation of intact organelles, and super-molecular structures where lysis conditions necessitate high impaction but low shearing.

  • Impact-resistant 2 mL tubes contain 1.6 mm aluminum oxide particles and 1.6 mm silicon carbide particles
  • With white caps
  • Specific Gravity: Aluminum Oxide, 3.95; Silicon Carbide, 3.20
  • Nucleic Acid Isolation and Purification
TRUSTED_SUSTAINABILITY

Specifications

Sample Type Bacteria (Gram +/-), Animal Tissues, Fungi, Plants, Coral Emulsions, Mold
DNase Dnase-free
For Use With (Application) Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis
Matrix Lysing matrix F
Molecular Formula Aluminum Oxide, Al2O3; Silicon Carbide, SiC
Quantity 50 x 2 mL Tubes
RNase RNase-free

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.