missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ LB-Agar, Powdered
Shop All MP Biomedicals Products
Click to view available options
Quantity:
454 g
1000 g
5000 g
10 x 500 mL
10 x 1000 mL
Form:
Powder
Content And Storage:
15°C to 30°C
Room Temperature
Unit Size:
10 L
1kg
454g
5 L
5kg
Specifications
Specifications
| Certifications/Compliance | Molecular biology Certified™ |
| Content And Storage | 15°C to 30°C |
| Form | Powder |
| Grade | Molecular biology |
| Product Type | Agar |
| Formulation | Tryptone 10g/L, Yeast extract 5g/L, NaCl 10g/L, Agar 15g/L |
| For Use With (Application) | For growth of bacteria |
| Quantity | 454 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction