missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Certifications/Compliance | Molecular Biology Certified™ |
| Content And Storage | 15°C to 30°C |
| Form | Powder |
| Grade | Molecular Biology |
| Product Type | Agar |
| Formulation | Tryptone 10g/L, Yeast extract 5g/L, Agar 15g/L |
| For Use With (Application) | For growth of bacteria |
| Quantity | 1000 g |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?