missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ LargeFlex™ Adapter for 2 x 250 mL Tube Holder on FastPrep 96

Product Code. 15252820 Shop All MP Biomedicals Products
Click to view available options
For Use With (Equipment):
FastPrep-96
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15252820

Brand: MP Biomedicals™ 116010590

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

LargeFlex™ Adapter on FastPrep 96 holds 2 x 250 mL Lysing Matrix Bottles. The ergonomic design of the adapter ensures an easy loading of Lysing Matrix bottles that remain securely sealed during the homogenization. For scale-up and small production run capabilities.

  • Product Overview: Super-sized adapter for the FastPrep-96™ that holds 2 x 250 mL Lysing Matrix Bottles. For scale-up and small production run capabilities. Compatible with the SafTest™ Food Inspection Platform. The ergonomic design of the adapter ensures an easy loading of Lysing Matrix bottles that remain securely sealed during the homogenization.
  • Additionally, frozen samples in Lysing Matrix tubes loaded in the adapter and stored at -20°C or -80°C are ready to be immediately processed with minimal hands-on manipulation, preventing degradation of cellular components by endogenous enzymes.
  • A wide range of disposable 250 mL Lysing Matrix bottles ensure thorough homogenization of any sample type in seconds.
  • Capacity: 2 tubes
TRUSTED_SUSTAINABILITY

Specifications

Product Type FastPrep-96 Adapters
For Use With (Application) Nucleic Acid Isolation and Purification
For Use With (Equipment) FastPrep-96

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.