missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Hydrolyzes Lactose (4-o-β-D-Galactopyranosyl-D-glucose) and produces β-D-Galactose and α-D-Glucose
- Catalyses transglycosylation of glucose into allolactose, the inducer of β-galactosidase
- EC Number: 232-864-1
- MDL Number: MFCD00130623
- Format: Off white powder
- Specific Activity: >5000U/g
- Stability: Stable in pH range of 4.0 to 8.0
- Stable to heat
- Soluble in water, insoluble in ethanol, acetone and isopropyl alcohol
- Optimum pH is 4.5 and 4.8 when O-Nitrophenyl-beta-D- galactopyranoside and Lactose are used as substrate
Used to screen for blue white colonies in MCS of various plasmid vectors in Escherichia coli or other bacteria and to break down lactose in milk to make it suitable for people with lactose intolerance.
Specifications
Specifications
| Concentration | >5000 U/g |
| Color | Off-White |
| pH | 4 to 8 |
| For Use With (Application) | Screen for blue white colonies in MCS of various plasmid vectors in E. coli or other Bacteria and to break down Lactose in milk |
| Content And Storage | 4°C |
| Source | Aspergillus oryzae |
| Quantity | 25 g |
| Product Type | Lactase |
| Form | Powder |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?