missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Nutritionally rich medium originally developed for growth and maintenance of E. coli strains
- Eliminates the waste, inaccuracies, and mess associated with weighing out bulk powder
- Contains essential electrolytes for transport and osmotic balance, due to NaCl component
Specifications
Specifications
| Content And Storage | 15°C to 30°C |
| Format | Capsules |
| Grade | Molecular Biology Certified™ |
| Media Type | Bacterial Media |
| Product Type | Microbiology Media |
| Sterility | Sterile |
| Formulation | 10g Tryptone, 5g Yeast Extract, 0.5g NaCl. Small quantities of biological buffers have been added for proper pH adjustment. |
| pH | 6.9 |
| Quantity | 454 g |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?