missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Kanamycin Sulfate
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Description
All antibiotics are intended for research use only. They are not for therapeutic use in humans or animals.
Antibiotic effective against most gram positive and gram negative bacteria and most types of yeast.
Specifications
Specifications
| Content And Storage | 15° to 30°C |
| Form | Crystalline powder |
| Product Type | Antibiotic |
| Quantity | 1 g |
Safety and Handling
Product Identifier
- Kanamycin Monosulfate, Cell Culture Reagent
- Danger
- Reproductive toxicity Category 1A
- H360-May damage fertility or the unborn child.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Kanamycin Monosulfate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction