missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Kanamycin Sulfate

Product Code. 11420782 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11420782

Brand: MP Biomedicals™ 0219453105

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Serve variety of purposes in cell culture, including selection, elimination of mycoplasma and microbial contamination, and prophylaxis

All antibiotics are intended for research use only. They are not for therapeutic use in humans or animals.

Antibiotic effective against most gram positive and gram negative bacteria and most types of yeast.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15° to 30°C
Product Type Antibiotic
Quantity 5 g
compliance-icons
Product Identifier
  • Kanamycin Monosulfate, Cell Culture Reagent
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1A
Hazard Statement
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Kanamycin Monosulfate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.