missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Serum, Type AB, Human, From Male Donors

Product Code. 11405055 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11405055 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11405055 Supplier MP Biomedicals™ Supplier No. 092930949

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as a growth supplement for cell culture media and in other research applications.

  • Collected from type AB human blood
  • Used as a growth supplement for cell culture media and in other research applications
  • Proven to grow many human cell lines at a faster rate than mixed blood group serum

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance U.S. regulations
Color Straw Yellow to Golden Brown
Content And Storage -20°C
Species Human
Form Liquid
Product Type Human Serum
Quantity 100 mL
Sterility Sterile
Sterilization Method 0.1 μm filtration
For Use With (Application) Grow many Human cell lines at a faster rate
Hemoglobin Concentration ≤26 mg/dL
Mycoplasma Detected No
Osmolality 263 to 323 mOsm/kg
pH 7 to 8
Protein Content 5 to 8.3 g/dL
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.