missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Human Albumin

Product Code. 11430412 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
11430412 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 11430412

Brand: MP Biomedicals™ 0219134901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

From human plasma

Negative for HBsAg and HIV antibodies as tested. Purity: >97%. Salt-free, lyophilized, homogeneous powder. MW: 65kDa.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) ELISA, Molecular Biology, Western Blot
Molecular Weight (g/mol) 65
Name Human Albumin
Quantity 1 g
Source Human
Storage Requirements 4°C
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.