missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Hormones: Dexamethasone

Product Code. 11454932 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mg
1000 mg
25 mg
500 mg
Unit Size:
100mg
1g
25mg
500mg
This item is not returnable. View return policy

Product Code. 11454932

Brand: MP Biomedicals™ 0219456125

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used to study apoptosis, cell signaling pathways and gene expression. MP Biomedicals Hormones: Dexamethasone acts as anti-inflammatory glucocorticoid that induces the production of phospholipase A2 inhibitory protein (lipocortin).

  • Inhibits induction of nitric oxide synthase (IC50=5 nM)
  • Causes reduction in cyclin A and Cdk2 activity, inhibition of G1/S transition in osteoblasts and inhibition of phosphorylation of Rb protein
  • Observed to induce apoptosis in human thymocytes and eosinophils but to inhibit apoptosis in neutrophils
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15° to 30°C
Form Crystalline powder
Product Type Dexamethasone
Quantity 25 mg
compliance-icons
Product Identifier
  • dexamethasone
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : dexamethasone

For research use only. Not intended for therapeutic use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.