missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Inhibits induction of nitric oxide synthase (IC50=5 nM)
- Causes reduction in cyclin A and Cdk2 activity, inhibition of G1/S transition in osteoblasts and inhibition of phosphorylation of Rb protein
- Observed to induce apoptosis in human thymocytes and eosinophils but to inhibit apoptosis in neutrophils
Specifications
Specifications
| Content And Storage | 15° to 30°C |
| Form | Crystalline powder |
| Product Type | Dexamethasone |
| Quantity | 1000 mg |
Safety and Handling
Product Identifier
- Dexamethasone, cell culture reagent
- Danger
- Reproductive toxicity Category 1B
- H360-May damage fertility or the unborn child.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: Dexamethasone
For research use only. Not intended for therapeutic use.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?