missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- No organic extractions are required, and up to 100 μg of DNA can be isolated per prep.
- The GNOME™ kit utilizes RNase Mixx to eliminate RNA immediately after lysis, and Protease Mixx to rapidly degrade cellular proteins. This is followed by a proprietary ''salting-out'' technique which precludes the need for phenol, chloroform, or other organic extractions.
- The DNA can either be spooled, removed with forceps, or centrifuged and dissolved in TE.
- The GNOME™ DNA Isolation kit is used to quickly and efficiently isolate high molecular weight genomic DNA from cells and tissues of any type, including bacteria, yeast, plant, and animal cells.
- Each preparation with the GNOME™ kit yields up to 100 μg of genomic DNA.
- The DNA isolated by the GNOME™ procedure is suitable for restriction enzyme digestion or PCR amplification in as little as 1 hour after cell lysis.
Specifications
Specifications
| Format | Kit |
| Sample Type | Blood, Tissues, Yeast, Bacteria, Plant |
| Includes | Cell Lysis/Denaturing Solution, RNase Mixx, Protease Mixx, Salt Out Mixture, Cell Suspension Solution |
| No. of Reactions | 100 Preps |
| pH | Cell Lysis/Denaturing Solution pH range: 6.2 to 6.9, Salt-out Mixture pH range: 6.0 to 6.8, Cell Suspension Solution pH range: 8.0 to 8.2 |
| Product Type | Gnome™ DNA Isolation Kit |
| Sample Volume | 20 to 50 mg wet weight bacterial cell pellet, Up to 250 mg plant tissue, 2 to 5 x 107 nucleated white blood cells, 50 to 250 mg mammalian tissue, ∼1 to 5 x 108 yeast cell pellet |
| Yield | 100 μg |
Safety and Handling
Product Identifier
- gnome DNA isolation kit
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- MIXTURE LIST-Contain: sodium dodecyl sulphate
Research Use Only (RUO). Ready for CE IVD labeling of clinical applications.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?