missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Gnome™ DNA Isolation Kit
Shop All MP Biomedicals Products
Click to view available options
No. of Reactions:
100 Preps
25 Preps
Unit Size:
1 set
100 preps
Description
- No organic extractions are required, and up to 100 μg of DNA can be isolated per prep.
- The GNOME™ kit utilizes RNase Mixx to eliminate RNA immediately after lysis, and Protease Mixx to rapidly degrade cellular proteins. This is followed by a proprietary ''salting-out'' technique which precludes the need for phenol, chloroform, or other organic extractions.
- The DNA can either be spooled, removed with forceps, or centrifuged and dissolved in TE.
- The GNOME™ DNA Isolation kit is used to quickly and efficiently isolate high molecular weight genomic DNA from cells and tissues of any type, including bacteria, yeast, plant, and animal cells.
- Each preparation with the GNOME™ kit yields up to 100 μg of genomic DNA.
- The DNA isolated by the GNOME™ procedure is suitable for restriction enzyme digestion or PCR amplification in as little as 1 hour after cell lysis.
Specifications
Specifications
| Format | Kit |
| Sample Type | Blood, Tissues, Yeast, Bacteria, Plant |
| Includes | Cell Lysis/Denaturing Solution, RNase Mixx, Protease Mixx, Salt Out Mixture, Cell Suspension Solution |
| No. of Reactions | 25 Preps |
| pH | Cell Lysis/Denaturing Solution pH range: 6.2 to 6.9, Salt-out Mixture pH range: 6.0 to 6.8, Cell Suspension Solution pH range: 8.0 to 8.2 |
| Product Type | Gnome™ DNA Isolation Kit |
| Sample Volume | 20 to 50 mg wet weight bacterial cell pellet, Up to 250 mg plant tissue, 2 to 5 x 107 nucleated white blood cells, 50 to 250 mg mammalian tissue, ∼1 to 5 x 108 yeast cell pellet |
| Yield | 100 μg |
Safety and Handling
Product Identifier
- gnome DNA isolation kit
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- EUH210-Safety data sheet available on request.
- MIXTURE LIST-Contains : sodium dodecyl sulphate
Research Use Only (RUO). Ready for CE IVD labeling of clinical applications.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction