missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ β-Glucosidase

Product Code. 11431102 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 kU
Unit Size:
25ku
This item is not returnable. View return policy

Product Code. 11431102

Brand: MP Biomedicals™ 0219519725

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

From Sweet Almonds

Activity: >1000U/mg material. Unit Definition: One unit is the amount of enzyme which liberates 1.0μg of glucose per minute at 35°C, pH 5.0. Salt-free. Lyophilized.

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥1000 U/mg
Color White
For Use With (Application) Synthesis of glucosides and fucosides
Content And Storage -20°C
Source Sweet almonds
Quantity 25 kU
Product Type beta-Glucosidase
Form Powder
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.