missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Gentamicin Sulfate
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
All antibiotics are intended for research use only. They are not for therapeutic use in humans or animals.
General antimicrobial effective against gram positive bacteria, gram negative bacteria, and mycoplasma.
Specifications
Specifications
| Content And Storage | 4°C |
| Form | Powder |
| Product Type | Antibiotic |
| Sterility | Non-sterile |
| pH | 3.5 to 5.5 |
| Quantity | 5 g |
Safety and Handling
Product Identifier
- Gentamicin Sulfate, Cell Culture Reagent
- Danger
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Gentamycin Sulfate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction