missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Geneclean™ Turbo Kit for PCR

Product Code. 11343129 Shop All MP Biomedicals Products
Change view
Click to view available options
No. of Reactions:
100 Preps
300 Preps
50 Preps
Unit Size:
100 preps
300 preps
50 preps
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
11343129 300 Preps 300 preps
11323129 50 Preps 50 preps
11333129 100 Preps 100 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11343129 Supplier MP Biomedicals™ Supplier No. 111103600

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Geneclean™ Turbo Kit optimized for PCR purification of DNA fragments of sizes 100 bp to 10 kb.

  • Ready for direct use
  • Removes primers, nucleotides, enzymes, mineral oil, salts, and other impurities
  • Applications: DNA Clean-up
TRUSTED_SUSTAINABILITY

Specifications

Sample Type Post amplification PCR reaction mixture (oil-free)
For Use With (Application) Isolation of DNA from PCR reactions, removal of primers, Nucleotides, Enzymes, Mineral Oil, salts and other impurities from DNA samples
Includes GeneClean™ Turbo™ Salt Solution, Wash Solution, Cartridges, Catch Tubes and Elution Solution
No. of Reactions 300 Preps
Sample Volume Up to 100 μL PCR Reaction Solution
Sample Source Gels and Solutions
compliance-icons compliance-icons
Product Identifier
  • GENECLEAN Turbo Salt Solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Guanidine Thiocyanate

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.