missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Geneclean™ Turbo Kit

Product Code. 11393119 Shop All MP Biomedicals Products
Change view
Click to view available options
No. of Reactions:
100 Preps
300 Preps
50 Preps
Unit Size:
1 set
300 preps
50 preps
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
11393119 50 Preps 50 preps
15122185 100 Preps 1 set
11313129 300 Preps 300 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11393119 Supplier MP Biomedicals™ Supplier No. 111102200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for purification of DNA fragments of sizes 0.1 kb to 300 kb from agarose gels, PCR reactions and other enzymatic solutions. MP Biomedicals™ Geneclean™ Turbo Kit can be used in enzymatic reactions, conventional or automated sequencing, cloning and microarray analysis.

  • Process with either vacuum or centrifugation and save time with patented Turbo filter design
  • Patented GeneClean™ technology simplifies the process of purifying DNA into three easy steps: Bind, Wash, and Elute
  • Ethanol precipitation is never required, and purified DNA is immediately ready for a wide variety of downstream applications
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room Temperature
Format Geneclean Imbedded Turbo Spin Column
For Use With (Application) Nucleic Acid isolation and purification
No. of Reactions 50 Preps
compliance-icons compliance-icons
Product Identifier
  • geneclean turbo kit
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: guanidine thiocyanate

Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.