missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- The GENECLEAN™ SPIN kit simplifies the process of purifying DNA into three easy steps: Bind, Wash and Elute
- Ethanol precipitation is never required and purified DNA is immediately ready for a wide variety of downstream applications
- Examples of applications are desalting, elimination of proteins from enzymatic reactions, removal of primers and unincorporated nucleotides from enzymatic reactions, separation of linearized from uncut vector and isolation of PCR products away from genomic DNA
Specifications
Specifications
| CAS | 593-84-0 |
| Format | Kit |
| Sample Type | Enzymatic Reactions, Solutions, Agarose Gels |
| For Use With (Application) | Isolate DNA from TAE or TBE buffered agarose gels and solutions |
| Includes | SPIN filters |
| No. of Reactions | 100 Preps |
| pH | Geneclean SPIN New Wash pH Range: 7.4 to 7.7, GC Spin Elution Solution pH Range: 6.8 to 7.2 |
| Product Type | Spin Kit |
| Sample Volume | Up to ∼300 mg Gel Slice or up to ∼300 μL DNA Solution |
| Synonym | DNA Clean-Up Kit |
| Show More |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?