missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Geneclean™ Glassmilk™ Silica Matrix

Product Code. 11393099 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1.5 mL
Unit Size:
1.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11393099 1.5 mL 1.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11393099 Supplier MP Biomedicals™ Supplier No. 111001404

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Geneclean™ Glassmilk™ Silica Matrix allows flexibility of reaction size and volume. Glassmilk™ slurry is a specially prepared aqeuous suspension of propietary silica matrix.

  • Geneclean™ Glassmilk™ is supplied with GeneClean™ Kit, which will bind to radioactive isotope-labeled ssDNA or dsDNA that is purified from agarose thus resulting in purified DNA for further use.

Specifications

CAS 593-84-0
Content And Storage Room Temperature
Format Kit
Isolation Technology Silica Matrix Technology
For Use With (Application) Nucleic acid purification
Product Type Glassmilk™ Slurry
Quantity 1.5 mL

Research Use Only (RUO). Ready for CE IVD labeling of clinical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.