missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ β- Galactosidase from Escherichia coli

Product Code. 11491792 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 kU
Unit Size:
1ku
This item is not returnable. View return policy

Product Code. 11491792

Brand: MP Biomedicals™ 0210493901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

β-galactosidase cleaves lactose into its monosaccharide components, glucose and galactose

  • CAS Number: 9031-11-2
  • EC Number: 232-864-1
  • Format: Pale gray fuffy powder
  • Specific activity: ≥50U/mg
  • Isoelectric point: 4.61
  • Optimum pH: 7 to 7.5

Used for structural investigation of carbohydrate, determination of lactose (foodstuff analysis) and as an enzyme label for enzyme immunoassay.

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥50 U/mg
Color Pale Gray
pH 7 to 7.5
For Use With (Application) Structural investigation of Carbohydrate, the determination of Lactose (food stuff analysis) and as an Enzyme label for Enzyme immunoassay
Content And Storage 4°C
Source E. coli
Quantity 1 kU
Product Type beta-Galactosidase
Molecular Weight (g/mol) 540000
Inhibitors p-Chloromercuribenzoate, lodoacetamide, Heavy Metal Ions (Zn++, Fe++, Zn++, Cd++, Cu++, Pb++, Ag+, Hg++), Ionic Detergents SDS, DAC
Form Lyophilized Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.