missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Ficin
Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Description
Activity: 300-500 milk clotting units/mg. Hygroscopic.
Specifications
Specifications
| Color | White |
| pH | 5 to 7.5 |
| For Use With (Application) | Cleavage of Mouse IgG |
| Content And Storage | 4°C |
| Source | Fig tree Ficus glabrata |
| Quantity | 5 g |
| Product Type | Ficin |
| Molecular Weight (g/mol) | ≈25 |
| Inhibitors | Iodoacetamide, Iodoacetic Acid, N-ethylmaleimide, Mercuric Chloride, DFP (Diisopropyl Fluorophosphate), TLCK (Na-p-Tosyl-lysine Chloromethyl Ketone), TPCK (N-Tosyl-L-phenylalanine Chloromethyl Ketone) |
| Form | Powder |
Safety and Handling
Product Identifier
- Ficin
- Danger
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Ficin
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction