missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Screw Caps

These 2 mL Screw Caps are for tubes to be used on FastPrep™ instruments or any bead beater.

Brand:  MP Biomedicals™ 115064002

Code : NEW

Additional Details : Weight : 0.17000kg

 View more versions of this product

Product Code. 11422340

  • £180.20 / Pack of 500
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Screw Caps with O-rings fit 2 mL screw cap tubes and are ideal for high-speed sample preparation instruments.
  • Screw Caps fit Empty Fastprep™ Tubes and are used in Fastprep systems, to hold sample for Homogenization.
  • Available separately for those who wish to fill their own Lysing Matrix Tubes.
  • Adapter Compatibility: ConeFlex Legacy (allows all FP24 adapters on FP96), CoolPrep (24x2 mL), HiPrep (48x2 mL), QuickFlex (96x2 mL), QuickPrep (24x2 mL), QuickPrep All Metal (24x2 mL)
  • Screw Caps can also be found in the FastDNA and FastDNA SPIN Kits.
Specifications

Specifications

2 mL
0.946 g/cm3
2 mL Lysing Matrix E Tubes
320°F (160°C)
500 Pk.
Purple
All
Nucleic Acid Isolation, Cell Culture, Proteomics, Purification and Detection, Protein Purification, DNA Purification, RNA Purification, Plant Tissue Processing, Tissue Analysis, Soil Analysis, Bacterial Culture, Yeast Culture, Fungal Tissue Analysis, Microbiome Studies
Screw Cap
Lysing Matrix E tubes have purple caps and are found in the Fast DNA Spin Kit for Soil and Fast RNA Pro Soil Kit.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.