missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ EMM w/o Dextrose, Powdered Media

Product Code. 11377859 Shop All MP Biomedicals Products
Click to view available options
:
227g
This item is not returnable. View return policy

Product Code. 11377859

Brand: MP Biomedicals™ 114110112

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Defined medium for the growth of S. pombe

Formulation

  • Use 12.35g powder per liter
  • Contents/liter: 3g phthalic acid K+, 2.2g Na2HPO4, 5g NH4Cl
  • Salts: 1.05g MgCl2·6H2O, 4.7mg CaCl2·2H2O, 1g KCl, 0.04g Na2SO4
  • Vitamins: 1mg pantothenic acid, 10mg nicotinic acid, 10mg myo-inositol, 1mg biotin
  • Minerals: 0.5mg boric acid, 0.4mg MnSO4, 0.4mg ZnSO4·7H2O, 0.2mg FeCl2·6H2O, 40μg molybdic acid, 0.1mg KI; 40μg CuSO4·5H20, 1mg citric acid

Yeast cell culture, Life sciences

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular biology Certified™
Content And Storage 15°C to 30°C
Form Powder
Grade Molecular biology
Product Type EMM
Formulation Potassium Hydrogen Phthalate 3g/L (14.7mM), Na2HPO4 2.2g/L (15.5mM), NH4Cl 5g/L (93.5mM), MgCl2.6H2O 1.05g/L (5.2mM), CaCl2.2H2O 14.7mg/L (99.8μM), KCl 1g/L (13.4mM), Na2SO4 0.04g/L (0.28mM), Pantothenic Acid 1mg/L (4.2μ
For Use With (Application) Used for the growth of Schizosaccharomyces pombe
Quantity 227 g
compliance-icons
Product Identifier
  • EMM w/o Dextrose (Powder)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Ammonium Chloride, Sodium Phosphate Dibasic Anhydrous, sodium sulphate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.