missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ DMEM/Ham's F-12 Medium without L-glutamine, sodium bicarbonate
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 x 10 L
Unit Size:
10 L
Description
- Acts as mammalian culture media
- Used as basal media for serum-free cell culture
Specifications
Specifications
| Form | Powder |
| Product Type | Dulbecco's Modified Eagle Medium |
| With Additives | Dextrose |
| Without Additives | No Glutamine, No Sodium Bicarbonate |
| For Use With (Application) | Mammalian cell culture applications |
| Quantity | 1 x 10 L |
| pH | 7.6 |
| Content And Storage | 0 to 5°C |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction