missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Dextrose

Product Code. 11412781 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
2.26 kg
11.33 kg
Unit Size:
25lb
5lb
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412781 2.26 kg 5lb
11422781 11.33 kg 25lb
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11412781 Supplier MP Biomedicals™ Supplier No. 0290559405
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Monohydrate

Dextrose monohydrate is a common natural sugar involved in processes such as energy production, glycosylation and formation of glycans that provide structure to cells. It is involved in a detrimental process in cells called glycation. It is used as a supplement for cell culture and in numerous cellular processes.

  • Formula:C6H12O6·H2O
  • Formula Weight: 198.18
  • CAS No.: 14431-43-7
TRUSTED_SUSTAINABILITY

Specifications

Product Type Animal Diet
Format Powder
For Use With (Application) Used as a supplement for cell culture and in numerous cellular processes
Quantity 2.26 kg
Storage Requirements 15°C to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.