missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ CSM-TRP-URA, Powdered Media
Dropout of Complete Supplement Mixture (CSM) of amino acids for S. cereviseae growth media
Brand: MP Biomedicals™ 114520512
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Formulation
- Add 0.72g of CSM-Trp-Ura to each liter of DOB or DOBA and autoclave at 121°C for 25 minutes for complete synthetic defined double drop-out medium
Applications:
Yeast cell culture, Gene expression, Transcription regulation
Specifications
Molecular biology Certified™ | |
Powder | |
Adenine 10mg/L, L-Arginine HCl 50mg/L, L-Aspartic Acid 80mg/L, L-Histidine HCl 20mg/L, L-Isoleucine 50mg/L, L-Leucine 100mg/L, L-Lysine HCl 50mg/L, L-Methionine 20mg/L, L-Phenylalanine 50mg/L, L-Threonine 100mg/L, L-Tryptophan 0mg/L, L-Tyrosine 50mg/L, Uracil 0mg/L, L-Valine 140mg/L | |
10 g |
15°C to 30°C | |
Yeast Media | |
For growth of Saccharomyces cerevisiae |