missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ CSM-HIS-LEU-TRP-URA, Powdered Media
Dropout supplement for S. cereviseae
Brand: MP Biomedicals™ 114540012
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Four dropout
- Supplement mixture of amino acids
- Complete formulations
- Dropout formulations
Formulation
- Add 0.60g of CSM-His-Leu-Trp-Ura to each liter of DOB or DOBA and autoclave at 121°C for 25 minutes for complete synthetic defined four dropout medium
Applications:
Yeast cell culture, Gene expression, Transcription
Specifications
Molecular biology Certified™ | |
Powder | |
Adenine 10mg/L, L-Arginine HCl 50mg/L, L-Aspartic Acid 80mg/L, L-Histidine HCl 0mg/L, L-Isoleucine 50mg/L, L-Leucine 0mg/L, L-Lysine HCl 50mg/L, L-Methionine 20mg/L, L-Phenylalanine 50mg/L, L-Threonine 100mg/L, L-Tryptophan 0mg/L, L-Tyrosine 50mg/L, Uracil 0mg/L, L-Valine 140mg/L | |
10 g |
15°C to 30°C | |
Yeast Media | |
For growth of Saccharomyces cerevisiae |