missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ CoolBigPrep™ Adapter for 2 x 50 mL Tube Holder on FastPrep-24

Product Code. 11471525 Shop All MP Biomedicals Products
Click to view available options
For Use With (Equipment):
FastPrep™- 24, FastPrep™-96
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11471525

Brand: MP Biomedicals™ 116002531

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

CoolBigPrep™ Adapter on FastPrep-24 holds 2 x 50 mL tubes for cryogenic cell grinding and lysis. It is used for extraction of sensitive biological compounds.

  • Product Overview: CoolBigPrep™ 2 x 50 mL Adapter is a cryogenic adapter which allows simultaneous lysis of 2 x 50 mL samples. Based on passive temperature control technology, the CoolBigPrep™ adapter ensures an efficient cooling of the samples as dry-ice, placed into the moving tray is in direct contact with the sample tubes. Due to a high heat transfer capacity and FastPrep™ precise settings of lysis parameters, the samples can be repeatably homogenized in their unthawed state. A wide range of disposable 50 mL Lysing Matrix tubes ensure thorough homogenization of any sample type in 40 seconds or less.
  • Applications: CoolBigPrep™ 2 x 50 mL Adapter is ideally suited for extractions of any temperature unstable or sensitive biological compounds such as RNA, siRNA, metabolites, intermediates, and enzymes from even the hardest samples to lyse.
  • Capacity: 2 tubes
  • Sample Volume: Up to 40 g
TRUSTED_SUSTAINABILITY

Specifications

Sample Size Up to 40 g
For Use With (Application) Ideal for extractions of temperature unstable biological compounds such as RNA, siRNA, metabolites, intermediates and enzymes
For Use With (Equipment) FastPrep™- 24, FastPrep™-96
Holds 2 x 50 mL Samples

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.