missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Collagenase from Clostridium histolyticum

Product Code. p-3179294 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 11470972

Brand: MP Biomedicals™ 02195109.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Enzyme responsible for break down of peptide bonds in collagen.

Assist in destroying extracellular structures in pathogenesis of bacteria such as Clostridium. These enzymes are exotoxins (a virulence factor) and help to facilitate the spread of gas gangrene. They normally target the connective tissue in muscle cells and other body organs. 

  • Synonyms: Clostridiopeptidase A
  • CAS Number: 9001-12-1
  • EC Number: 232-582-9
  • MDL Number: MFCD00130830
  • Format: Dialyzed, lyophilized brown lumpy powder
  • Specific Activity: >125U/mg
  • Solubility: Dissolve enzyme at concentration of 1mg/mL in 0.05M TES with 0.36mM calcium chloride, pH 7.5 

Note: This item is for research use only.  It is not a compendial item and is not meant for medical use.

TRUSTED_SUSTAINABILITY

Specifications

Concentration >125 U/mg
Color Brown
pH 7 to 9
Content And Storage 4°C
Source Clostridium histolyticum
Quantity 100 mg
Product Type Collagenase
Molecular Weight (g/mol) 105000 and 57400
Inhibitors Metal chelating agents such as Cysteine, EDTA or o-Phenanthroline
Form Lyophilized Powder
compliance-icons compliance-icons
Product Identifier
  • Collagenase, Type I
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : collagenase
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.