missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Collagenase from Clostridium histolyticum
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Assist in destroying extracellular structures in pathogenesis of bacteria such as Clostridium. These enzymes are exotoxins (a virulence factor) and help to facilitate the spread of gas gangrene. They normally target the connective tissue in muscle cells and other body organs.
- Synonyms: Clostridiopeptidase A
- CAS Number: 9001-12-1
- EC Number: 232-582-9
- MDL Number: MFCD00130830
- Format: Dialyzed, lyophilized brown lumpy powder
- Specific Activity: >125U/mg
- Solubility: Dissolve enzyme at concentration of 1mg/mL in 0.05M TES with 0.36mM calcium chloride, pH 7.5
Note: This item is for research use only. It is not a compendial item and is not meant for medical use.
Specifications
Specifications
| Concentration | >125 U/mg |
| Color | Brown |
| pH | 7 to 9 |
| Content And Storage | 4°C |
| Source | Clostridium histolyticum |
| Quantity | 100 mg |
| Product Type | Collagenase |
| Molecular Weight (g/mol) | 105000 and 57400 |
| Inhibitors | Metal chelating agents such as Cysteine, EDTA or o-Phenanthroline |
| Form | Lyophilized Powder |
Safety and Handling
Product Identifier
- Collagenase, Type I
- Danger
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : collagenase
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction