missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Chorionic Gonadotropin (hCG)

Product Code. 11492721 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
This item is not returnable. View return policy

Product Code. 11492721

Brand: MP Biomedicals™ 0219859110

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

From human pregnancy urine

Lyophilized.

TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) ∽39500
Name Chorionic Gonadotropin from Human Pregnancy Urine
Quantity 10 mg
Source Human Pregnancy Urine
Storage Requirements -20°C
Form Lyophilized
compliance-icons
Product Identifier
  • Chorionic Gonadotropin, USP
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Supplemental information
  • MIXTURE LIST-Contains : Chorionic Gonadotropin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.