missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Chorionic Gonadotropin (hCG)
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
Description
Lyophilized.
Specifications
Specifications
| Molecular Weight (g/mol) | ∽39500 |
| Name | Chorionic Gonadotropin from Human Pregnancy Urine |
| Quantity | 100 mg |
| Source | Human Pregnancy Urine |
| Storage Requirements | -20°C |
| Form | Lyophilized |
Safety and Handling
Product Identifier
- Chorionic Gonadotropin, USP
- Warning
- Reproductive toxicity Category 2
- H361-Suspected of damaging fertility or the unborn child.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Chorionic Gonadotropin
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction