missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Choline Oxidase from Alcaligones Species

Product Code. 11498771 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 U
1000 U
500 U
Unit Size:
100 units
1ku
500 units
This item is not returnable. View return policy

Product Code. 11498771

Brand: MP Biomedicals™ 0215349280

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

>10 units/mg solid, Yellowish powder. For cholinesterase activity assays.

TRUSTED_SUSTAINABILITY

Specifications

Concentration >10 U/mg
Color Yellow
pH 8 to 8.5
For Use With (Application) Enzymatic determination of Phospholipids by coupling with Phospholipase D and for Cholinesterase activity assays
Content And Storage -20°C
Source Alcaligenes species
Quantity 100 U
Product Type Choline Oxidase
Molecular Weight (g/mol) ≈95000
Inhibitors p-Chloromercuribenzoate, Cu++, Co++, Hg++, Ag+
Form Lyophilized Powder
Show More Show Less
compliance-icons
Product Identifier
  • Choline Oxidase
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Choline Oxidase
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.