missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Cholesterol Esterase from Pseudomonas Species

Product Code. 11431892 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kU
200 U
Unit Size:
1ku
200 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11431892 1 kU 1ku
11421892 200 U 200 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11431892 Supplier MP Biomedicals™ Supplier No. 0210543991

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cholesterol esterase (CE) is a reversible enzyme that can hydrolyze or synthesize fatty acid esters of cholesterol and other sterols

  • CAS Number: 9026-00-0
  • EC Number: 232-808-6
  • MDL Number: MFCD00071052
  • Format: yellowish tan lyophilized powder
  • Specific activity: ≥100U/mg (solid)
  • Isoelectric point: 5.9 ±0.1
  • Stable at -20°C for at least one year

Cholesterol Esterase is useful for enzymatic determination of total cholesterol when coupled with cholesterol oxidase in clinical analysis.

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥100 U/mg
Color Yellowish Tan
pH 7 to 9
For Use With (Application) Enzymatic determination of Total Cholesterol when coupled with Cholesterol Oxidase in clinical analysis
Content And Storage 0°C
Source Pseudomonas species
Quantity 1 kU
Product Type Cholesterol Esterase
Molecular Weight (g/mol) ≈300000
Inhibitors Hg- - Ag ionic detergents
Form Lyophilized Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.