missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Cellulase from Aspergillus niger

Product Code. 11467881 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
This item is not returnable. View return policy

Product Code. 11467881

Brand: MP Biomedicals™ 0215058380

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cellulase from Aspergillus niger catalyzes hydrolysis of endo-1,4-β-D-glycosidic linkages in cellulose, lichenin, barley glucan and cellooligosaccharides cellotriose to cellohexaose

Cellulase refers to a group of enzymes which acting together hydrolyze cellulose. The b-glucosidases, including cellobiose are primarily active on the smaller molecular weight cellulose hydrolysates.

  • CAS Number: 9012-54-8
  • EC Number: 232-734-4
  • MDL Number: MFCD00081510
  • Format: Tan powder
  • Specific activity: >60000U/g
  • Readily soluble in water
  • Foreign activity: Salmonella Negative
  • Optimum pH: 4.2 to 5.2

Cellulase is used in molecular biology, cell biology, textile industry, pulp and paper industry and for commercial food processing in coffee

TRUSTED_SUSTAINABILITY

Specifications

Concentration >60,000 U/g
Color Tan
pH 4.2 to 5.2
For Use With (Application) Cellulase is used for commercial food processing in coffee, textile industry and in laundry detergents, pharmaceutical applications, fermentation of biomass into biofuels
Content And Storage 4°C
Source Aspergillis niger
Quantity 100 g
Product Type Cellulase
Form Powder
compliance-icons
Product Identifier
  • Cellulase
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : cellulase
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.