missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Catalase

Product Code. p-3179290 Shop All MP Biomedicals Products
Click to view available options
Quantity:
50 mL
100 mL
Unit Size:
100mL
50mL
This item is not returnable. View return policy

Product Code. 11463070

Brand: MP Biomedicals™ 0210040250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fungal suspension from Aspergillus niger

Catalase, [9001-05-2]

Density: 1.2g/mL
Activity: 1000 units/ml

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥1000 U/mL
For Use With (Application) Removal of Peroxides, the generation of Oxygen and in coupled systems, for the determination of metabolites
Content And Storage 4°C
Source Aspergillus niger
Quantity 50 mL
Product Type Catalase
Molecular Weight (g/mol) ≈250
Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.