missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Catalase
Shop All MP Biomedicals Products
Click to view available options
Quantity:
50 mL
100 mL
Unit Size:
100mL
50mL
Description
Catalase, [9001-05-2]
Density: 1.2g/mL
Activity: 1000 units/ml
Specifications
Specifications
| Concentration | ≥1000 U/mL |
| For Use With (Application) | Removal of Peroxides, the generation of Oxygen and in coupled systems, for the determination of metabolites |
| Content And Storage | 4°C |
| Source | Aspergillus niger |
| Quantity | 100 mL |
| Product Type | Catalase |
| Molecular Weight (g/mol) | ≈250 |
| Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction