missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ BigFlex™ Adapter for 8 x 50 mL Tube Holder for FastPrep 96
Shop All MP Biomedicals Products
Click to view available options
For Use With (Equipment):
FastPrep-96
Unit Size:
Each
Description
- Product Overview: BigFlex™ 8 x 50 mL Adapter for FastPrep-96™ is a large-scale adapter for the FastPrep-96™ that holds 8 x 50 mL Lysing Matrix tubes (compatible with SafTest™ Food Inspection System). A wide range of disposable 50 mL Lysing Matrix tubes ensure thorough homogenization of any sample type in seconds.
- Application: BigFlex™ 8 x 50 mL Adapter for FastPrep-96™ is a large sample volume adapter is ideally suited for DNA and RNA isolation, enzyme isolation and protein production, natural products isolation, food preparation for quality analysis, biopharma manufacturing, and forensic applications. Additionally, frozen samples in Lysing Matrix tubes loaded in the adapter and stored at -20°C or -80°C are ready to be immediately processed with minimal hands-on manipulation, preventing degradation of cellular components by endogenous enzymes.
- Capacity: 8 tubes
- Sample Volume: Up to 160 g
Specifications
Specifications
| Sample Volume (Metric) | Up to 160 g |
| For Use With (Application) | To grind or homogenize samples at ambient temperature |
| For Use With (Equipment) | FastPrep-96 |
| Holds | 8 x 50 mL Tubes |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction