missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Antibiotics: Puromycin Dihydrochloride
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
Description
- Format: Powder, Non-sterile
All antibiotics are intended for research use only. They are not for therapeutic use in humans or animals.
Specifications
Specifications
| Content And Storage | -20°C |
| Form | Powder |
| Product Type | Antibiotic |
| Quantity | 100 mg |
Safety and Handling
Product Identifier
- Puromycin Dihydrochloride, cell culture reagent
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Puromycin Dihydrochloride
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction