missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Format: Powder, Non-sterile
All antibiotics are intended for research use only. They are not for therapeutic use in humans or animals.
Specifications
Specifications
| Content And Storage | -20°C |
| Form | Powder |
| Product Type | Antibiotic |
| Quantity | 10 mg |
Safety and Handling
Product Identifier
- Puromycin Dihydrochloride, cell culture reagent
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: Puromycin Dihydrochloride
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?