missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Alcohol Oxidase
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 kU
5 kU
Unit Size:
1ku
5ku
Description
Catalyzes the oxidation of an alcohol to an aldehyde and hydrogen peroxide. Activity: 10-20U per mg protein. Unit Definition: One unit will oxidize 1.0μmole of methanol to formaldehyde per minute at pH 7.5 at 25°C. Solution in sucrose.
Specifications
Specifications
| Concentration | ≥10 U/mg |
| Color | Reddish Brown |
| pH | 7.2 |
| For Use With (Application) | Colorimetric determination of Blood Alcohol |
| Storage Buffer | 30% Sucrose, 0.1M Potassium Phosphate Buffer at pH 8 |
| Content And Storage | -20°C |
| Source | Pichea pastoris |
| Quantity | 1 kU |
| Product Type | Alcohol Oxidase |
| Molecular Weight (g/mol) | ≈630000 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction