missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Alcohol Oxidase

Product Code. 11484602 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 kU
5 kU
Unit Size:
1ku
5ku
This item is not returnable. View return policy

Product Code. 11484602

Brand: MP Biomedicals™ 0219015501

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

From Pichea pastoris E.C.1.1.3.13

Catalyzes the oxidation of an alcohol to an aldehyde and hydrogen peroxide. Activity: 10-20U per mg protein. Unit Definition: One unit will oxidize 1.0μmole of methanol to formaldehyde per minute at pH 7.5 at 25°C. Solution in sucrose.

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥10 U/mg
Color Reddish Brown
pH 7.2
For Use With (Application) Colorimetric determination of Blood Alcohol
Storage Buffer 30% Sucrose, 0.1M Potassium Phosphate Buffer at pH 8
Content And Storage -20°C
Source Pichea pastoris
Quantity 1 kU
Product Type Alcohol Oxidase
Molecular Weight (g/mol) ≈630000
Form Frozen
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.